This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00477043_g1
See other PPIC GE Assays ›
Gene Symbol:
Ppic
Gene Name:
peptidylprolyl isomerase C
Gene Aliases:
CyP-20c
Chromosome Location:
Chr.18: 53406341 - 53418007 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00477043_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppic
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008908.4 NP_032934.1 2 - 3 287 56
GenBank mRNA AK131931.1 - 2 - 3 295 56
AK150654.1 - 2 - 3 253 56
AK151449.1 - 2 - 3 253 56
AK168911.1 - 2 - 3 271 56
BC025861.1 - 2 - 3 302 5026674 56
M74227.1 - 2 - 3 287 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001338.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppic

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
peptidyl-prolyl cis-trans isomerase activity
protein binding
cyclosporin A binding
isomerase activity
peptide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800009843.mm.1 NM_008908.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800009843.mm.1 NM_008908.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800009843.mm.1 TC1800001338.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800009844.mm.1 NM_008908.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800009844.mm.1 NM_008908.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800009844.mm.1 TC1800001338.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0