This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00477785_g1
See other NFE2L2 GE Assays ›
Gene Symbol:
Nfe2l2
Gene Name:
nuclear factor, erythroid derived 2, like 2
Gene Aliases:
AI194320, Nrf2
Chromosome Location:
Chr.2: 75675519 - 75704641 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00477785_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nfe2l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010902.3 NP_035032.1 2 - 3 545 146
GenBank mRNA AK142347.1 - 2 - 3 447 146
AK144247.1 - 2 - 3 419 146
BC003340.1 - 2 - 3 401 146
BC026943.1 - 2 - 3 486 3663276 146
U20532.1 - 2 - 3 561 146

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003791.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nfe2l2

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
inflammatory response
proteasomal ubiquitin-independent protein catabolic process
positive regulation of gene expression
protein ubiquitination
positive regulation of blood coagulation
endoplasmic reticulum unfolded protein response
cellular response to oxidative stress
response to endoplasmic reticulum stress
cellular response to drug
positive regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of transcription from RNA polymerase II promoter in response to oxidative stress
PERK-mediated unfolded protein response
cellular response to glucose starvation
proteasome-mediated ubiquitin-dependent protein catabolic process
cell redox homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of embryonic development
negative regulation of cell death
cellular response to hydrogen peroxide
cellular response to tumor necrosis factor
cellular response to fluid shear stress
cellular response to laminar fluid shear stress
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
negative regulation of hydrogen peroxide-induced cell death
positive regulation of glutathione biosynthetic process
regulation of removal of superoxide radicals
negative regulation of endothelial cell apoptotic process
positive regulation of reactive oxygen species metabolic process
transcription regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II activating transcription factor binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcription cofactor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein domain specific binding
sequence-specific DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200032404.mm.1 NM_010902.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200032404.mm.1 TC0200003791.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline