This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00478045_m1
See other PDE1C GE Assays ›
Gene Symbol:
Pde1c
Gene Name:
phosphodiesterase 1C
Gene Aliases:
-
Chromosome Location:
Chr.6: 56069798 - 56652516 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00478045_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025568.2 NP_001020739.1 3 - 4 790 78
NM_001159952.1 NP_001153424.1 2 - 3 763 78
NM_001159953.1 NP_001153425.1 3 - 4 790 78
NM_001159955.1 NP_001153427.1 4 - 5 352 78
NM_001159956.1 NP_001153428.1 2 - 3 209 78
NM_001159957.1 NP_001153429.1 3 - 4 236 78
NM_001159960.1 NP_001153432.1 4 - 5 664 78
NM_011054.4 NP_035184.1 4 - 5 767 78
GenBank mRNA AK014887.1 - 2 - 3 180 78
AK015375.1 - 4 - 5 321 78
AK017183.1 - 3 - 4 774 78
AK029531.1 - 3 - 4 234 78
AK030423.1 - 3 - 4 792 78
AK050499.1 - 4 - 5 665 78
AK077123.1 - 2 - 3 158 78
AK082180.1 - 3 - 4 777 78
AK082497.1 - 3 - 4 575 78
AK133243.1 - 4 - 5 336 78
AK139923.1 - 2 - 3 737 78
BB614593.1 - 2 - 3 210 78
BC032277.1 - 3 - 4 719 5400689 78
L76944.1 - 3 - 4 420 78
L76946.1 - 4 - 5 766 78
L76947.1 - 3 - 4 253 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002331.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde1c

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
sensory perception of smell
cGMP catabolic process
negative regulation of insulin secretion
response to calcium ion
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
calmodulin binding
phosphoric diester hydrolase activity
hydrolase activity
cAMP binding
metal ion binding
calcium- and calmodulin-regulated 3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001025568.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001025568.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159952.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159952.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159953.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159953.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159955.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159955.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159956.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159956.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159957.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159957.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159960.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_001159960.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_011054.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 NM_011054.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600018232.mm.1 TC0600002331.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline