This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00478255_g1
See other PLD3 GE Assays ›
Gene Symbol:
Pld3
Gene Name:
phospholipase D family, member 3
Gene Aliases:
Sam-9
Chromosome Location:
Chr.7: 27531645 - 27553213 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00478255_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pld3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011116.1 NP_035246.1 3 - 4 351 59
GenBank mRNA AF026124.1 - 3 - 4 351 59
AK153508.1 - 1 - 2 40 59
BC068170.1 - 3 - 4 348 59
BC076586.1 - 3 - 4 415 30604186 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002669.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF026124.1,AK153508.1,BC076586.1,NM_011116.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pld3

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
catalytic activity
phospholipase D activity
hydrolase activity
N-acylphosphatidylethanolamine-specific phospholipase D activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700023312.mm.1 TC0700002669.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700023312.mm.1 NM_011116.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700023309.mm.1 TC0700002669.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700023309.mm.1 NM_011116.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700023316.mm.1 TC0700002669.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700023319.mm.1 TC0700002669.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0