This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00478438_g1
See other PTPN18 GE Assays ›
Gene Symbol:
Ptpn18
Gene Name:
protein tyrosine phosphatase, non-receptor type 18
Gene Aliases:
FLP1, HSCF, PTP-HSCF, PTP-K1, Ptpk1
Chromosome Location:
Chr.1: 34459746 - 34473784 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00478438_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011206.2 NP_035336.2 11 - 12 998 97
GenBank mRNA AK144353.1 - 11 - 12 980 97
BC008512.1 - 11 - 12 968 3591418 97
CD740968.1 - 4 - 5 97
U35124.1 - 11 - 12 972 97
U49853.1 - 11 - 12 938 97
U52523.1 - 11 - 12 986 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000224.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn18

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
peptidyl-tyrosine dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
protein binding
hydrolase activity
phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100001450.mm.1 NM_011206.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001450.mm.1 NM_011206.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100001450.mm.1 TC0100000224.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0