This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00478652_g1
See other ASH2L GE Assays ›
Gene Symbol:
Ash2l
Gene Name:
ash2 (absent, small, or homeotic)-like (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.8: 25816000 - 25847694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00478652_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ash2l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080793.2 NP_001074262.1 3 - 4 328 69
NM_001286207.1 NP_001273136.1 3 - 4 301 69
NM_011791.3 NP_035921.2 3 - 4 418 69
GenBank mRNA AB020983.2 - 3 - 4 400 69
AK087934.1 - 3 - 4 393 69
AK132471.1 - 3 - 4 400 69
AK145576.1 - 3 - 4 293 69
AK145790.1 - 3 - 4 305 69
AK145967.1 - 3 - 4 272 69
AK146254.1 - 3 - 4 330 69
AK146599.1 - 3 - 4 302 69
AK146756.1 - 3 - 4 293 69
AK146938.1 - 3 - 4 395 69
BC012957.1 - 3 - 4 405 4211433 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001923.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ash2l

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
positive regulation of cell proliferation
chromatin modification
response to estrogen
positive regulation of transcription from RNA polymerase II promoter
chromatin-mediated maintenance of transcription
histone H3-K4 methylation
DNA binding
protein binding
histone methyltransferase activity (H3-K4 specific)
transcription regulatory region DNA binding
metal ion binding
euchromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800015358.mm.1 NM_001080793.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800015358.mm.1 NM_001286207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800015358.mm.1 NM_011791.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800015358.mm.1 TC0800001923.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lzcpb:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0