This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479036_m1
See other PHTF1 GE Assays ›
Gene Symbol:
Phtf1
Gene Name:
putative homeodomain transcription factor 1
Gene Aliases:
AU041898, AW049785, Phft, Phtf
Chromosome Location:
Chr.3: 103968110 - 104007490 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479036_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phtf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163467.1 NP_001156939.1 8 - 9 940 89
NM_001163468.1 NP_001156940.1 7 - 8 1187 89
NM_001163469.1 NP_001156941.1 7 - 8 1211 89
NM_013629.2 NP_038657.2 8 - 9 1346 89
GenBank mRNA AJ133721.1 - 8 - 9 940 89
AJ242864.1 - 8 - 9 1346 89
AK019146.1 - 8 - 9 1250 89
AK088714.1 - 7 - 8 1078 89
AK131629.1 - 1237 89
BC021147.1 - 8 - 9 1219 89
BC050825.1 - 7 - 8 1023 6518647 89
BC057340.1 - 8 - 9 1232 6827816 89
BC060177.1 - 8 - 9 1232 6827816 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001070.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phtf1

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 NM_001163467.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 NM_001163467.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 NM_001163468.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 NM_001163468.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 NM_001163469.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 NM_001163469.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 NM_013629.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 NM_013629.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008681.mm.1 TC0300001070.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 NM_001163467.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 NM_001163467.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 NM_001163468.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 NM_001163468.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 NM_001163469.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 NM_001163469.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 NM_013629.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 NM_013629.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008682.mm.1 TC0300001070.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0