This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479058_g1
See other PTPN5 GE Assays ›
Gene Symbol:
Ptpn5
Gene Name:
protein tyrosine phosphatase, non-receptor type 5
Gene Aliases:
Step
Chromosome Location:
Chr.7: 47077795 - 47133695 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479058_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163565.1 NP_001157037.1 3 - 4 592 104
NM_013643.2 NP_038671.2 3 - 4 700 104
GenBank mRNA AK038416.1 - 3 - 4 369 104
BC051975.1 - 3 - 4 341 104
BC079592.1 - 3 - 4 672 6817360 104
U28216.1 - 3 - 4 453 104
U28217.1 - 3 - 4 711 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003050.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn5

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
positive regulation of receptor internalization
protein dephosphorylation
positive regulation of neuron projection development
dephosphorylation
positive regulation of protein import into nucleus, translocation
peptidyl-tyrosine dephosphorylation
exploration behavior
response to immobilization stress
positive regulation of long-term synaptic potentiation
positive regulation of neuron death
positive regulation of protein targeting to mitochondrion
positive regulation of response to drug
phosphotyrosine binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
phosphatase activity
protein kinase binding
glutamate receptor binding
mitogen-activated protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700027917.mm.1 NM_001163565.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700027917.mm.1 NM_001163565.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700027917.mm.1 NM_013643.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700027917.mm.1 NM_013643.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700027917.mm.1 TC0700003050.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0