This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479731_m1
See other PNLIPRP1 GE Assays ›
Gene Symbol:
Pnliprp1
Gene Name:
pancreatic lipase related protein 1
Gene Aliases:
Plrp1
Chromosome Location:
Chr.19: 58728887 - 58744169 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479731_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnliprp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018874.2 NP_061362.1 2 - 3 202 70
GenBank mRNA AF061274.1 - 2 - 3 203 70
AK028105.1 - 2 - 3 115 70
BC090985.1 - 2 - 3 114 30315884 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000841.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pnliprp1

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
triglyceride lipase activity
lipase activity
metal ion binding
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007298.mm.1 NM_018874.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007298.mm.1 NM_018874.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007298.mm.1 TC1900000841.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900007300.mm.1 NM_018874.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007300.mm.1 NM_018874.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007300.mm.1 TC1900000841.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900007299.mm.1 NM_018874.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007299.mm.1 NM_018874.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007299.mm.1 TC1900000841.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900007297.mm.1 NM_018874.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007297.mm.1 NM_018874.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007297.mm.1 TC1900000841.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0