This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479736_g1
See other PNLIPRP1 GE Assays ›
Gene Symbol:
Pnliprp1
Gene Name:
pancreatic lipase related protein 1
Gene Aliases:
Plrp1
Chromosome Location:
Chr.19: 58728887 - 58744169 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479736_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnliprp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018874.2 NP_061362.1 7 - 8 850 58
GenBank mRNA AF061274.1 - 7 - 8 851 58
AK028105.1 - 7 - 8 763 58
BC068266.1 - 3 - 4 406 58
BC090985.1 - 7 - 8 762 30315884 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000841.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pnliprp1

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
triglyceride lipase activity
lipase activity
metal ion binding
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007305.mm.1 NM_018874.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007305.mm.1 NM_018874.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007305.mm.1 TC1900000841.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0