This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479833_m1
See other ATF7IP GE Assays ›
Gene Symbol:
Atf7ip
Gene Name:
activating transcription factor 7 interacting protein
Gene Aliases:
2610204M12Rik, AM, Mcaf1
Chromosome Location:
Chr.6: 136518789 - 136610850 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479833_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atf7ip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019426.2 NP_062299.2 8 - 9 2381 78
GenBank mRNA AJ132702.1 - 8 - 9 2374 78
AK132359.1 - 7 - 8 1001 78
AK167542.1 - 9 - 10 2427 78
BC037617.1 - 2383 78
BC052460.1 - 8 - 9 2381 6834550 78
BC060246.1 - 8 - 9 2364 5705945 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001622.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atf7ip

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA methylation
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
regulation of RNA polymerase II transcriptional preinitiation complex assembly
transcription corepressor activity
ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600012921.mm.1 TC0600001622.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600012921.mm.1 NM_019426.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600012921.mm.1 NM_019426.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-r9tfj:80/100.66.76.159:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline