This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479955_g1
See other PPP4C GE Assays ›
Gene Symbol:
Ppp4c
Gene Name:
protein phosphatase 4, catalytic subunit
Gene Aliases:
1110002D08Rik, AU016079, Ppx
Chromosome Location:
Chr.7: 126785866 - 126792471 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479955_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp4c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019674.3 NP_062648.1 3 - 4 257 108
GenBank mRNA AF088911.1 - 3 - 4 282 108
AK144244.1 - 2 - 3 184 108
BC001993.1 - 3 - 4 257 3491568 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004295.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp4c

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

regulation of double-strand break repair via homologous recombination
dephosphorylation
signal transduction by protein phosphorylation
NIK/NF-kappaB signaling
NF-kappaB-inducing kinase activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
protein binding
lamin binding
hydrolase activity
phosphatase activity
protein complex binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700035939.mm.1 NM_019674.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035939.mm.1 NM_019674.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700035939.mm.1 TC0700004295.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tx8s7:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0