This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479960_g1
See other PPP4C GE Assays ›
Gene Symbol:
Ppp4c
Gene Name:
protein phosphatase 4, catalytic subunit
Gene Aliases:
1110002D08Rik, AU016079, Ppx
Chromosome Location:
Chr.7: 126785866 - 126792471 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479960_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp4c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019674.3 NP_062648.1 8 - 9 909 73
GenBank mRNA AF088911.1 - 8 - 9 934 73
AK027923.1 - 6 - 7 689 73
AK144244.1 - 7 - 8 836 73
AK199030.1 - 1 - 2 136 73
BC001993.1 - 8 - 9 909 3491568 73
U79747.1 - 4 - 5 561 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004295.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp4c

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

regulation of double-strand break repair via homologous recombination
dephosphorylation
signal transduction by protein phosphorylation
NIK/NF-kappaB signaling
NF-kappaB-inducing kinase activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
protein binding
lamin binding
hydrolase activity
phosphatase activity
protein complex binding
metal ion binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502254 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502251 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502258 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502257 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502255 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502256 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502253 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 AK027923.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 AK144244.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 BC001993.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 NM_019674.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 AF088911.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 AK027923.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 AK144244.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 BC001993.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 NM_019674.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P275408 AF088911.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608343 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596461 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377737 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281485 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836372 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155474 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 AK027923.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 AK144244.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 BC001993.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 NM_019674.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 AF088911.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 AK027923.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 AK144244.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 BC001993.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 NM_019674.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P275408 AF088911.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502269 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502254 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502267 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502249 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502251 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502244 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502272 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502253 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502243 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502268 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502270 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502263 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502255 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502247 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502245 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502241 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502261 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502273 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502265 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502258 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502256 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01630300 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155474 - -
Whole Mouse Genome G4122A A_52_P275408 AK027923.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 AK144244.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 BC001993.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 NM_019674.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 AF088911.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 AK027923.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 AK144244.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 BC001993.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 NM_019674.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P275408 AF088911.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208394 U79747.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208394 AK027923.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208394 AK144244.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208394 AK199030.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208394 BC001993.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139547 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155474 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06207153 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06207151 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06626404 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06626398 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502256 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502257 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502255 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502251 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502253 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502258 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25502254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502174 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502256 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502232 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502287 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502228 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502150 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502160 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502183 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502310 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502270 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502255 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502211 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502238 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502188 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502204 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502213 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502244 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502173 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502241 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502298 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502314 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502245 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502190 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502258 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502203 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502209 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502208 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502229 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502206 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502317 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502269 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502229 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502263 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502237 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06626403 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06385797 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06130581 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502264 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502268 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06209092 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502228 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502233 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502269 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502272 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05927623 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502224 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502261 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502232 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502236 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06310973 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502249 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502251 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502244 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06328974 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502221 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502220 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502230 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502258 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06524534 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502213 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502209 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06385781 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502270 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502218 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06626398 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502241 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502211 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502214 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502225 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502259 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502267 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25502253 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10568047 - -
Affymetrix Mouse Gene 1.0 ST Array 10568048 - -
Affymetrix Mouse Gene 1.0 ST Array 10568045 - -
Affymetrix Mouse Gene 1.0 ST Array 10568043 - -
Affymetrix Mouse Gene 1.0 ST Array 10568044 - -
Affymetrix Mouse Gene 1.0 ST Array 10568049 - -
Affymetrix Mouse Gene 1.0 ST Array 10568041 - -
Affymetrix Mouse Gene 1.0 ST Array 10568042 - -
Affymetrix Mouse Gene 1.0 ST Array 10568046 - -
Affymetrix HT MG-430 PM Array Plate 1460288_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10568040 - -
Mouse Genome 430 2.0 Array 1445350_at - -
Mouse Genome 430 2.0 Array 1445350_PM_at - -
Mouse Genome 430 2.0 Array 1460288_a_at - -
Mouse Genome 430 2.0 Array 1460288_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 56420_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 56420_at - -
Affymetrix Mouse Clariom S Array PSR0700035933.mm.1 NM_019674.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035933.mm.1 NM_019674.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700035933.mm.1 TC0700004295.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700035934.mm.1 NM_019674.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035934.mm.1 NM_019674.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700035934.mm.1 TC0700004295.mm.1 MTA TC
Mouse Expression Set 430 1460288_a_at - -
Mouse Expression Set 430 1460288_PM_a_at - -
Murine Genome U74v2 Set 95631_PM_at - -
Murine Genome U74v2 Set 95631_at - -
Mouse Genome 430A 2.0 Array 1460288_PM_a_at - -
Mouse Genome 430A 2.0 Array 1460288_a_at - -
Affymetrix HT Mouse Genome 430A Array 1460288_a_at - -
Affymetrix HT Mouse Genome 430A Array 1460288_PM_a_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 5130593 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005130593 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237696 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1237696 - -
mouse-6 v1.1 (Illumina) scl30672.9.1_29-S - -
Illumina Sentrix mouse-6.v1 bead array 5130593 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl30672.9.1_29-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130593 - -
MouseRef-8 v2 Expression BeadChip ILMN_1237696 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237696 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl30672.9.1_29-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0