This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00480321_m1
See other IGDCC4 GE Assays ›
Gene Symbol:
Igdcc4
Gene Name:
immunoglobulin superfamily, DCC subclass, member 4
Gene Aliases:
DDM36, Nope
Chromosome Location:
Chr.9: 65101461 - 65137947 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00480321_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igdcc4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290315.1 NP_001277244.1 5 - 6 885 64
NM_020043.3 NP_064427.2 5 - 6 885 64
GenBank mRNA AB052620.1 - 5 - 6 885 64
AB052621.1 - 5 - 6 885 64
AF176694.1 - 5 - 6 836 64
AK079073.1 - 5 - 6 851 64
AK143256.1 - 5 - 6 837 64
CD352400.1 - 4 - 5 522 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000829.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igdcc4

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900006295.mm.1 NM_020043.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006295.mm.1 TC0900000829.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006296.mm.1 NM_020043.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006296.mm.1 TC0900000829.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline