This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00480614_m1
See other RBFOX1 GE Assays ›
Gene Symbol:
Rbfox1
Gene Name:
RNA binding protein, fox-1 homolog (C. elegans) 1
Gene Aliases:
A2bp, A2bp1, Hrnbp1, fox-1
Chromosome Location:
Chr.16: 5884793 - 7412480 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00480614_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbfox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021477.5 NP_067452.2 11 - 12 2004 61
NM_183188.2 NP_899011.2 7 - 8 899 61
GenBank mRNA AB041596.2 - 10 - 11 1311 61
AF107204.1 - 7 - 8 687 61
AF191501.1 - 7 - 8 900 61
AY659954.1 - 7 - 8 675 61
AY659955.1 - 7 - 8 675 61
AY659956.1 - 7 - 8 735 61
AY659957.1 - 7 - 8 735 61
AY659958.1 - 6 - 7 642 61
BC059002.1 - 10 - 11 1668 6821627 61
EF410136.1 - 9 - 10 1055 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000054.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbfox1

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
regulation of RNA splicing
neuromuscular process controlling balance
nucleotide binding
nucleic acid binding
RNA binding
protein C-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600000509.mm.1 NM_021477.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000509.mm.1 NM_021477.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600000509.mm.1 NM_183188.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000509.mm.1 NM_183188.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000509.mm.1 TC1600000054.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0