This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00480692_m1
See other METTL9 GE Assays ›
Gene Symbol:
Mettl9
Gene Name:
methyltransferase like 9
Gene Aliases:
0610012D09Rik, AA517660, Drev, MNCb-5680
Chromosome Location:
Chr.7: 121032794 - 121076835 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00480692_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mettl9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021554.2 NP_067529.2 2 - 3 719 114
GenBank mRNA AB041664.1 - 2 - 3 497 114
AJ278576.1 - 2 - 3 719 114
AK075586.1 - 2 - 3 482 114
AK169951.1 - 1 - 2 135 114
AK201397.1 - 2 - 3 208 115
BC023188.1 - 1 - 2 160 114
BC068124.1 - 2 - 3 513 5716972 114
BC082317.1 - 1 - 2 144 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001739.mm.1

Additional Information:

This assay is in the 5'UTR region of the AB041664.1,BC023188.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mettl9

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700014802.mm.1 NM_021554.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014802.mm.1 NM_021554.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700014802.mm.1 TC0700001739.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 843786 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline