This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00480941_m1
See other PPP1R7 GE Assays ›
Gene Symbol:
Ppp1r7
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 7
Gene Aliases:
2310014J01Rik, SDS22
Chromosome Location:
Chr.1: 93343645 - 93367619 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00480941_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023200.2 NP_075689.1 1 - 2 64 100
GenBank mRNA AF222867.1 - 1 - 2 64 100
AK009870.1 - 1 - 2 57 100
AK031983.1 - 856 100
AK129024.1 - 1 - 2 81 100
AK145136.1 - 1 - 2 70 100
AK162517.1 - 1 - 2 72 100
BC013524.1 - 1 - 2 65 4237290 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000929.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF222867.1,AK009870.1,AK145136.1,AK162517.1,BC013524.1,NM_023200.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r7

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase modulator

Gene Ontology Categories:

Function(s)

chromosome segregation
positive regulation of protein dephosphorylation

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100007888.mm.1 NM_023200.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007888.mm.1 NM_023200.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100007888.mm.1 TC0100000929.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100007890.mm.1 NM_023200.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007890.mm.1 NM_023200.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100007890.mm.1 TC0100000929.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 392509 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline