This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00480945_m1
See other PPP1R7 GE Assays ›
Gene Symbol:
Ppp1r7
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 7
Gene Aliases:
2310014J01Rik, SDS22
Chromosome Location:
Chr.1: 93343645 - 93367619 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00480945_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023200.2 NP_075689.1 5 - 6 451 124
GenBank mRNA AF222867.1 - 5 - 6 451 124
AK009342.1 - 5 - 6 466 124
AK009870.1 - 5 - 6 444 124
AK031983.1 - 1243 124
AK129024.1 - 5 - 6 468 124
AK145136.1 - 5 - 6 457 124
AK162517.1 - 5 - 6 459 124
BC013524.1 - 5 - 6 452 4237290 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000929.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r7

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase modulator

Gene Ontology Categories:

Function(s)

chromosome segregation
positive regulation of protein dephosphorylation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100007895.mm.1 NM_023200.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007895.mm.1 NM_023200.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100007895.mm.1 TC0100000929.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100007896.mm.1 NM_023200.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007896.mm.1 NM_023200.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100007896.mm.1 TC0100000929.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline