This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00481309_m1
See other OGFOD3 GE Assays ›
Gene Symbol:
Ogfod3
Gene Name:
2-oxoglutarate and iron-dependent oxygenase domain containing 3
Gene Aliases:
1110031I02Rik
Chromosome Location:
Chr.11: 121177593 - 121204648 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00481309_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogfod3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025402.2 NP_079678.1 7 - 8 775 78
GenBank mRNA AK004005.1 - 7 - 8 776 78
AK145938.1 - 7 - 8 840 78
AK168300.1 - 7 - 8 811 78
BC024349.1 - 7 - 8 742 4972349 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004248.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogfod3

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
sulfonate dioxygenase activity
molecular_function
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
L-ascorbic acid binding
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100040715.mm.1 NM_025402.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040715.mm.1 NM_025402.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100040715.mm.1 TC1100004248.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0