| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700002731.mm.1 |
NM_025425.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002731.mm.1 |
NM_025425.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002731.mm.1 |
TC1700000350.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002730.mm.1 |
NM_025425.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002730.mm.1 |
NM_025425.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002730.mm.1 |
TC1700000350.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002733.mm.1 |
NM_001163945.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002733.mm.1 |
NM_001163945.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002733.mm.1 |
NM_025425.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002733.mm.1 |
NM_025425.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002733.mm.1 |
TC1700000350.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700002732.mm.1 |
NM_001163945.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002732.mm.1 |
NM_001163945.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002732.mm.1 |
NM_025425.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002732.mm.1 |
NM_025425.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700002732.mm.1 |
TC1700000350.mm.1 |
MTA TC |