This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00481379_g1
See other SSR2 GE Assays ›
Gene Symbol:
Ssr2
Gene Name:
signal sequence receptor, beta
Gene Aliases:
1500032E05Rik, AI315033, AU020133, TLAP, TRAPB, TRAPbeta
Chromosome Location:
Chr.3: 88579584 - 88588420 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00481379_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK005340.1 - 1 - 2 77 65
AK010565.1 - 1 - 2 44 65
AK020145.1 - 1 - 2 27 65
AK084530.1 - 1 - 2 44 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK005340.1, AK010565.1, AK020145.1, AK084530.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 915133 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0