This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00481383_m1
See other SSR2 GE Assays ›
Gene Symbol:
Ssr2
Gene Name:
signal sequence receptor, beta
Gene Aliases:
1500032E05Rik, AI315033, AU020133, TLAP, TRAPB, TRAPbeta
Chromosome Location:
Chr.3: 88579584 - 88588420 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00481383_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025448.3 NP_079724.1 4 - 5 656 77
GenBank mRNA AK005340.1 - 5 - 6 527 77
AK007538.1 - 2 - 3 123 77
AK007720.1 - 2 - 3 123 77
AK010565.1 - 5 - 6 494 77
AK012281.1 - 4 - 5 484 77
AK020145.1 - 5 - 6 477 77
AK050505.1 - 4 - 5 657 77
AK084530.1 - 5 - 6 494 77
AK214352.1 - 2 - 3 204 77
AK217251.1 - 1 - 2 39 77
BC010214.1 - 4 - 5 640 3497894 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000731.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssr2

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314615 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314618 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314617 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314616 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK012281.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK020145.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK050505.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK084530.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK213548.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK012281.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK020145.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK050505.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK084530.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314264 AK213548.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138251 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584182 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262128 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518852 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12251 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111599 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK012281.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK020145.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK050505.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK084530.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK213548.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK012281.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK020145.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK050505.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK084530.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314264 AK213548.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK012281.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK020145.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK050505.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK084530.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK213548.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK012281.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK020145.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK050505.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK084530.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314264 AK213548.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314597 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314594 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314629 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314628 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314611 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314608 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314623 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314625 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314607 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314630 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314615 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314613 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314618 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314596 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314621 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111599 - -
Whole Mouse Genome G4122A A_51_P314264 AK012281.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK020145.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK050505.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK084530.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK213548.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK012281.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK020145.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK050505.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK084530.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P314264 AK213548.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103333 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111599 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02912769 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02363512 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02363511 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314615 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314616 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314618 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314682 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314666 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314669 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314695 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314659 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314685 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314681 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314664 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314592 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314698 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314662 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314712 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314690 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314651 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02413105 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314653 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314601 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161104 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02305096 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314623 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314694 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314677 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161083 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02811716 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161106 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161099 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314673 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02743105 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314590 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03049923 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161098 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314654 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161087 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02852033 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314649 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161096 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314695 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314589 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161091 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314607 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314681 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02813321 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161089 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02363514 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314666 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314634 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03049924 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314675 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02530162 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02912769 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314669 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314646 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161107 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314704 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314706 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314648 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02813324 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314628 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161085 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314640 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314624 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02399404 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02363512 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02392743 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02782832 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02363516 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02392746 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03161088 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314595 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02363507 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03145760 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03162798 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314705 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314636 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22314629 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10493266 - -
Affymetrix Mouse Gene 1.0 ST Array 10493263 - -
Affymetrix Mouse Gene 1.0 ST Array 10493261 - -
Affymetrix Mouse Gene 1.0 ST Array 10493260 - -
Affymetrix Mouse Gene 1.0 ST Array 10493264 - -
Affymetrix Mouse Gene 1.0 ST Array 10493265 - -
Affymetrix Mouse Gene 1.0 ST Array 10493262 - -
Affymetrix HT MG-430 PM Array Plate 1449930_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10493259 - -
Mouse Genome 430 2.0 Array 1449930_PM_a_at - -
Mouse Genome 430 2.0 Array 1449930_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66256_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66256_at - -
Affymetrix Mouse Clariom S Array PSR0300005177.mm.1 NM_025448.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300005177.mm.1 NM_025448.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300005177.mm.1 TC0300000731.mm.1 MTA TC
Mouse Expression Set 430 1449930_PM_a_at - -
Mouse Expression Set 430 1449930_a_at - -
Murine Genome U74v2 Set 101061_at - -
Murine Genome U74v2 Set 101061_PM_at - -
Mouse Genome 430A 2.0 Array 1449930_PM_a_at - -
Mouse Genome 430A 2.0 Array 1449930_a_at - -
Affymetrix HT Mouse Genome 430A Array 1449930_a_at - -
Affymetrix HT Mouse Genome 430A Array 1449930_PM_a_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 5390180 - -
Illumina mouse-6 v1.0 expression beadchip 5390180 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005390180 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2626538 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2626538 - -
mouse-6 v1.1 (Illumina) scl22994.6_49-S - -
Illumina Sentrix mouse-6.v1 bead array 5390180 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl22994.6_49-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390180 - -
MouseRef-8 v2 Expression BeadChip ILMN_2833606 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2833606 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22994.6_49-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline