This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00481432_g1
See other SMIM8 GE Assays ›
Gene Symbol:
Smim8
Gene Name:
small integral membrane protein 8
Gene Aliases:
1810030N24Rik, 2810406B13Rik
Chromosome Location:
Chr.4: 34768664 - 34778396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00481432_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smim8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025471.2 NP_079747.1 2 - 3 293 67
GenBank mRNA AK005941.1 - 2 - 3 296 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002391.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK005941.1,NM_025471.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smim8

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400019418.mm.1 NM_025471.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400019418.mm.1 TC0400002391.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400019419.mm.1 NM_025471.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400019419.mm.1 TC0400002391.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline