This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00481622_m1
See other 1700029F12RIK GE Assays ›
Gene Symbol:
1700029F12Rik
Gene Name:
RIKEN cDNA 1700029F12 gene
Gene Aliases:
-
Chromosome Location:
Chr.13: 97021864 - 97035254 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00481622_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700029F12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025585.3 NP_079861.1 2 - 3 398 67
NR_104276.1 2 - 3 260 67
GenBank mRNA AK006036.1 - 2 - 3 386 67
AK006488.1 - 2 - 3 366 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002475.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene 1700029F12Rik

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248566 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248569 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248558 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248561 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248565 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248562 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248568 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 AK006036.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 AK006488.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 BC049576.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 NM_001080777.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 NM_025585.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 AK006036.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 AK006488.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 BC049576.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 NM_001080777.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P568968 NM_025585.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907980 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36226 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498069 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84170 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 AK006036.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 AK006488.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 BC049576.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 NM_001080777.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 NM_025585.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 AK006036.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 AK006488.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 BC049576.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 NM_001080777.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P568968 NM_025585.2 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07963928 - -
Whole Mouse Genome G4122A A_52_P430411 AK006036.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P430411 AK006488.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P430411 BC049576.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P430411 BU962717.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P430411 NM_001080777.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P466068 AK006488.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P466068 NM_025585.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 AK006036.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 AK006488.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 BC049576.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 NM_001080777.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 NM_025585.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 AK006036.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 AK006488.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 BC049576.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 NM_001080777.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P568968 NM_025585.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100320 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07963928 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248568 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248566 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248565 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248562 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248561 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248569 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29248558 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29249074 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29249081 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29249107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29249060 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29249068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29249066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29249076 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29249110 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29249061 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10773317 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29249107 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29249112 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29249060 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10411357 - -
Affymetrix Mouse Gene 1.0 ST Array 10411356 - -
Affymetrix Mouse Gene 1.0 ST Array 10411358 - -
Affymetrix Mouse Gene 1.0 ST Array 10411355 - -
Affymetrix Mouse Gene 1.0 ST Array 10411354 - -
Affymetrix HT MG-430 PM Array Plate 1419715_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10411353 - -
Mouse Genome 430 2.0 Array 1419715_at - -
Mouse Genome 430 2.0 Array 1419715_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66479_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66479_at - -
Affymetrix Mouse Clariom S Array PSR1300016617.mm.1 NM_001080777.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300016617.mm.1 NM_001284205.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016617.mm.1 NM_025585.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300016617.mm.1 NR_104276.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016617.mm.1 TC1300002475.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300016618.mm.1 NM_001080777.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300016618.mm.1 NM_001284205.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016618.mm.1 NM_025585.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300016618.mm.1 NR_104276.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016618.mm.1 TC1300002475.mm.1 MTA TC
Mouse Expression Set 430 1419715_PM_at - -
Mouse Expression Set 430 1419715_at - -
Murine Genome U74v2 Set 168874_i_at - -
Murine Genome U74v2 Set 168874_PM_i_at - -
Mouse Genome 430A 2.0 Array 1419715_at - -
Mouse Genome 430A 2.0 Array 1419715_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1419715_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1419715_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 370344 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 4150341 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004150341 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218314 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1218314 - -
mouse-6 v1.1 (Illumina) scl0066479.1_162-S - -
Illumina Sentrix mouse-6.v1 bead array 4150341 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0066479.1_162-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150341 - -
MouseRef-8 v2 Expression BeadChip ILMN_3116055 - -
MouseRef-8 v2 Expression BeadChip ILMN_1218314 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3116055 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218314 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0