This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00481821_m1
See other TRAPPC5 GE Assays ›
Gene Symbol:
Trappc5
Gene Name:
trafficking protein particle complex 5
Gene Aliases:
4021401A16Rik, TRS31
Chromosome Location:
Chr.8: 3676477 - 3680921 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00481821_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trappc5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025701.4 NP_079977.3 1 - 2 127 71
GenBank mRNA AK003633.1 - 1 - 2 95 71
AK007989.1 - 1 - 2 70
AK076212.1 - 1 - 2 128 71
AK078451.1 - 1 - 2 93 71
BC049179.1 - 1 - 2 85 6313540 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000022.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK003633.1,AK007989.1,AK076212.1,AK078451.1,BC049179.1,NM_025701.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trappc5

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
vesicle-mediated transport
protein binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615115 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615118 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615132 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615117 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615129 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615128 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615130 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615116 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 BC049179.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 AK078451.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 AK171833.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 BC028946.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 NM_025701.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 BC049179.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 AK078451.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 AK171833.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 BC028946.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P137691 NM_025701.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 BC049179.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 AK078451.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 AK171833.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 BC028946.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 NM_025701.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 BC049179.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 AK078451.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 AK171833.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 BC028946.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P208377 NM_025701.4 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903134 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793243 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157110 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48707 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108246 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 BC049179.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 AK078451.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 AK171833.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 BC028946.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 NM_025701.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 BC049179.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 AK078451.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 AK171833.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 BC028946.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P208377 NM_025701.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 BC049179.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 AK078451.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 AK171833.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 BC028946.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 NM_025701.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 BC049179.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 AK078451.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 AK171833.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 BC028946.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P137691 NM_025701.4 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615113 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615123 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615108 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615106 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615103 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615091 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615109 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615098 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615120 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615100 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615104 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615097 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615117 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615110 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615127 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615118 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615101 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615121 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615119 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615096 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615094 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615126 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615114 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01662861 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108246 - -
Whole Mouse Genome G4122A A_52_P137691 BC049179.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 AK078451.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 AK171833.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 BC028946.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 NM_025701.4 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 BC049179.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 AK078451.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 AK171833.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 BC028946.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P137691 NM_025701.4 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 BC049179.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 AK078451.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 AK171833.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 BC028946.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 NM_025701.4 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 BC049179.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 AK078451.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 AK171833.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 BC028946.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P208377 NM_025701.4 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122125 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108246 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07374665 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07374666 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07374674 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07220334 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615128 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615117 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615129 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615116 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615132 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615115 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615118 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25615130 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615207 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615194 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615098 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615074 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615127 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615072 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615096 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615104 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615188 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615080 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615071 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615136 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615117 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615101 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615108 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615106 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615205 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615200 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615069 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615176 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615203 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615081 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615120 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615103 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07100124 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615121 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615114 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06805815 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615132 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07252507 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615096 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615108 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07100126 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615117 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615106 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615136 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615123 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615094 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615141 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615087 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07100119 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615124 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615120 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615101 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615098 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615090 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615110 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07148265 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06805826 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615086 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615138 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07374668 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07220334 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615109 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615140 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25615104 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10569888 - -
Affymetrix Mouse Gene 1.0 ST Array 10569889 - -
Affymetrix Mouse Gene 1.0 ST Array 10569887 - -
Affymetrix HT MG-430 PM Array Plate 1418233_PM_a_at - -
Affymetrix HT MG-430 PM Array Plate 1448999_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10569886 - -
Mouse Genome 430 2.0 Array 1418233_a_at - -
Mouse Genome 430 2.0 Array 1418233_PM_a_at - -
Mouse Genome 430 2.0 Array 1448999_PM_at - -
Mouse Genome 430 2.0 Array 1448999_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66682_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66682_PM_at - -
Affymetrix Mouse Clariom S Array PSR0800000286.mm.1 NM_025701.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800000286.mm.1 NM_025701.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800000286.mm.1 TC0800000022.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800000285.mm.1 NM_025701.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800000285.mm.1 NM_025701.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800000285.mm.1 TC0800000022.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800000284.mm.1 NM_025701.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800000284.mm.1 NM_025701.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800000284.mm.1 TC0800000022.mm.1 MTA TC
Mouse Expression Set 430 1448999_at - -
Mouse Expression Set 430 1448999_PM_at - -
Mouse Expression Set 430 1418233_a_at - -
Mouse Expression Set 430 1418233_PM_a_at - -
Murine Genome U74v2 Set 94554_at - -
Murine Genome U74v2 Set 94554_PM_at - -
Mouse Genome 430A 2.0 Array 1448999_PM_at - -
Mouse Genome 430A 2.0 Array 1418233_PM_a_at - -
Mouse Genome 430A 2.0 Array 1448999_at - -
Mouse Genome 430A 2.0 Array 1418233_a_at - -
Affymetrix HT Mouse Genome 430A Array 1418233_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1418233_a_at - -
Affymetrix HT Mouse Genome 430A Array 1448999_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1448999_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 485619 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2320315 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002320315 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2475271 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2475271 - -
mouse-6 v1.1 (Illumina) scl34125.2_74-S - -
Illumina Sentrix mouse-6.v1 bead array 2320315 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl34125.2_74-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320315 - -
MouseRef-8 v2 Expression BeadChip ILMN_2845600 - -
MouseRef-8 v2 Expression BeadChip ILMN_2475271 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2475271 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2845600 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl34125.2_74-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0