This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00482048_m1
See other HACD4 GE Assays ›
Gene Symbol:
Hacd4
Gene Name:
3-hydroxyacyl-CoA dehydratase 4
Gene Aliases:
4933428I03Rik, Hcad4, Ptplad2
Chromosome Location:
Chr.4: 88407687 - 88438935 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00482048_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hacd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025760.4 NP_080036.1 1 - 2 92 118
GenBank mRNA AK016968.1 - 1 - 2 76 118
AK017257.1 - 1 - 2 95 118
AK039344.1 - 82 118
AK039602.1 - 82 118
AK048282.1 - 1 - 2 86 118
AK050927.1 - 1 - 2 86 118
BC020155.1 - 1 - 2 78 4016383 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003000.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK016968.1,AK017257.1,AK039344.1,AK039602.1,AK048282.1,AK050927.1,BC020155.1,NM_025760.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hacd4

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
fatty acid elongation
very long-chain fatty acid biosynthetic process
lyase activity
3-hydroxyacyl-CoA dehydratase activity
enzyme binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400024287.mm.1 TC0400003000.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400024287.mm.1 NM_025760.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024287.mm.1 NM_025760.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400024286.mm.1 TC0400003000.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400024286.mm.1 NM_025760.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024286.mm.1 NM_025760.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400024285.mm.1 TC0400003000.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400024285.mm.1 NM_025760.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400024285.mm.1 NM_025760.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 594065 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0