This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00482316_m1
See other TRAPPC13 GE Assays ›
Gene Symbol:
Trappc13
Gene Name:
trafficking protein particle complex 13
Gene Aliases:
2410002O22Rik, 2610524F24Rik, AI451892, AU017229
Chromosome Location:
Chr.13: 104142153 - 104178703 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00482316_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trappc13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001093759.1 NP_001087228.1 2 - 3 419 133
NM_001093760.1 NP_001087229.1 2 - 3 419 133
NM_025879.2 NP_080155.3 2 - 3 419 133
GenBank mRNA AK010346.1 - 2 - 3 400 133
AK013956.1 - 2 - 3 377 133
AK084373.1 - 2 - 3 369 133
AK149832.1 - 2 - 3 133
AK163342.1 - 2 - 3 407 133
AK167749.1 - 2 - 3 379 133
BC021756.1 - 2 - 3 360 3584010 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002596.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK010346.1,AK013956.1,AK084373.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trappc13

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017681.mm.1 NM_001093759.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017681.mm.1 NM_001093759.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017681.mm.1 NM_001093760.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017681.mm.1 NM_001093760.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017681.mm.1 NM_025879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017681.mm.1 NM_025879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300017681.mm.1 TC1300002596.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0