This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00482370_m1
See other POLR3K GE Assays ›
Gene Symbol:
Polr3k
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide K
Gene Aliases:
12.3kDa, 1500004O14Rik, AI196849, AU014893, C11, RPC10, RPC11
Chromosome Location:
Chr.2: 181864360 - 181870830 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00482370_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr3k
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025901.3 NP_080177.1 1 - 2 212 75
GenBank mRNA AK005152.1 - 1 - 2 116 75
AK008637.1 - 1 - 2 84 75
AK010875.1 - 1 - 2 150 75
AK021353.1 - 1 - 2 147 75
AK031110.1 - 1 - 2 147 75
AK088206.1 - 1 - 2 112 75
BC025898.1 - 1 - 2 228 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002837.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr3k

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

immune system process
transcription, DNA-templated
termination of RNA polymerase III transcription
innate immune response
defense response to virus
RNA polymerase III activity
nucleic acid binding
DNA binding
DNA-directed RNA polymerase activity
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200023561.mm.1 NM_025901.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023561.mm.1 NM_025901.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200023561.mm.1 TC0200002837.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200023560.mm.1 NM_025901.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023560.mm.1 NM_025901.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200023560.mm.1 TC0200002837.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 856164 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0