This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00482776_g1
See other GTF3C6 GE Assays ›
Gene Symbol:
Gtf3c6
Gene Name:
general transcription factor IIIC, polypeptide 6, alpha
Gene Aliases:
2410016F19Rik, AU019813
Chromosome Location:
Chr.10: 40247701 - 40258158 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00482776_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf3c6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026113.4 NP_080389.2 3 - 4 232 82
GenBank mRNA AK005261.1 - 3 - 4 231 82
AK007816.1 - 3 - 4 233 82
AK010514.1 - 3 - 4 232 82
AK162053.1 - 3 - 4 232 82
AK168070.1 - 3 - 4 231 82
BC048446.1 - 3 - 4 238 6590885 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002042.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf3c6

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase III promoter
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000014499.mm.1 NM_026113.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014499.mm.1 NM_026113.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000014499.mm.1 TC1000002042.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0