This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00482789_m1
See other UNC50 GE Assays ›
Gene Symbol:
Unc50
Gene Name:
unc-50 homolog (C. elegans)
Gene Aliases:
1110002A21Rik, GMH1, UNCL, URP
Chromosome Location:
Chr.1: 37430144 - 37439124 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00482789_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc50
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026123.3 NP_080399.1 5 - 6 968 100
GenBank mRNA AK003273.1 - 5 - 6 743 100
AK004433.1 - 5 - 6 732 100
AK132757.1 - 5 - 6 969 100
AK135885.1 - 5 - 6 960 100
AK210936.1 - 2 - 3 146 100
AK215289.1 - 1 - 2 71 100
AY017214.1 - 5 - 6 953 100
BC019484.1 - 5 - 6 935 4039903 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000260.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc50

Gene Ontology Categories:

Function(s) Process(es)

transport
cell surface receptor signaling pathway
protein transport
RNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153986 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153984 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153985 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153987 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026794 AY017214.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026794 BC019484.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026794 AK004433.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026794 AK132757.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026794 AK003273.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335374 BC052409.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923542 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562134 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313389 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 705457 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14405 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149455 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026794 AY017214.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026794 BC019484.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026794 AK004433.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026794 AK132757.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026794 AK003273.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153988 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153995 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154011 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154017 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153980 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154008 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154010 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153977 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153981 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154021 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154004 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153999 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153987 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154016 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153997 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154015 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154019 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153982 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153991 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154005 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154006 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154012 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153993 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153978 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154001 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153986 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153983 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153979 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20154013 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153996 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149455 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00046498 - -
Whole Mouse Genome G4122A A_51_P474509 AY017214.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474509 BC019484.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474509 AK004433.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474509 AK132757.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P474509 AK003273.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135381 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149455 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01145221 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00350292 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00350285 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153984 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153985 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153986 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154049 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154013 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153996 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154043 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154057 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154010 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154034 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154055 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153993 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153991 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153983 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154015 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153999 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154051 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154059 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154045 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154036 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154006 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20154005 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154051 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154047 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154043 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154012 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154008 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154049 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154041 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00256055 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154055 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00256032 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00350285 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00256028 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154033 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154062 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154057 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00256058 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01059838 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00256052 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00256061 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01145221 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154039 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154059 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20153997 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00256043 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154036 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01059844 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154004 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154011 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01059866 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154013 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154056 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154032 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00256062 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154034 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00350298 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154019 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154030 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00350290 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154061 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20154015 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00949248 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20153993 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10345611 - -
Affymetrix Mouse Gene 1.0 ST Array 10345613 - -
Affymetrix Mouse Gene 1.0 ST Array 10345612 - -
Affymetrix Mouse Gene 1.0 ST Array 10345614 - -
Affymetrix Mouse Gene 1.0 ST Array 10345615 - -
Affymetrix Mouse Gene 1.0 ST Array 10345610 - -
Affymetrix Mouse Gene 1.0 ST Array 10345609 - -
Affymetrix HT MG-430 PM Array Plate 1421982_PM_a_at - -
Affymetrix HT MG-430 PM Array Plate 1435811_PM_a_at - -
Affymetrix HT MG-430 PM Array Plate 1435812_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10345608 - -
Mouse Genome 430 2.0 Array 1435812_at - -
Mouse Genome 430 2.0 Array 1435811_PM_a_at - -
Mouse Genome 430 2.0 Array 1435812_PM_at - -
Mouse Genome 430 2.0 Array 1421982_a_at - -
Mouse Genome 430 2.0 Array 1435811_a_at - -
Mouse Genome 430 2.0 Array 1421982_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67387_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67387_PM_at - -
Affymetrix Mouse Clariom S Array PSR0100001865.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001865.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001865.mm.1 TC0100000260.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001862.mm.1 NM_026123.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001862.mm.1 NM_026123.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001862.mm.1 TC0100000260.mm.1 MTA TC
Mouse Expression Set 430 1435812_PM_at - -
Mouse Expression Set 430 1435811_PM_a_at - -
Mouse Expression Set 430 1435812_at - -
Mouse Expression Set 430 1421982_a_at - -
Mouse Expression Set 430 1421982_PM_a_at - -
Mouse Expression Set 430 1435811_a_at - -
Murine Genome U74v2 Set 96675_at - -
Murine Genome U74v2 Set 96675_PM_at - -
Mouse Genome 430A 2.0 Array 1435811_PM_a_at - -
Mouse Genome 430A 2.0 Array 1421982_a_at - -
Mouse Genome 430A 2.0 Array 1421982_PM_a_at - -
Mouse Genome 430A 2.0 Array 1435812_PM_at - -
Mouse Genome 430A 2.0 Array 1435811_a_at - -
Mouse Genome 430A 2.0 Array 1435812_at - -
Affymetrix HT Mouse Genome 430A Array 1435812_at - -
Affymetrix HT Mouse Genome 430A Array 1435811_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1435812_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1421982_a_at - -
Affymetrix HT Mouse Genome 430A Array 1435811_a_at - -
Affymetrix HT Mouse Genome 430A Array 1421982_PM_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 654696 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4920037 - -
Illumina mouse-6 v1.0 expression beadchip 4920037 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004920037 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2457194 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2457194 - -
mouse-6 v1.1 (Illumina) scl067387.5_4-S - -
Illumina Sentrix mouse-6.v1 bead array 4920037 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl067387.5_4-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920037 - -
MouseRef-8 v2 Expression BeadChip ILMN_2457194 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2457194 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0