This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00482894_m1
See other APOBEC1 GE Assays ›
Gene Symbol:
Apobec1
Gene Name:
apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1
Gene Aliases:
Cdar1
Chromosome Location:
Chr.6: 122577792 - 122603041 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00482894_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apobec1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134391.1 NP_001127863.1 4 - 5 515 63
NM_031159.3 NP_112436.1 4 - 5 571 63
GenBank mRNA AK146427.1 - 4 - 5 447 63
AK151799.1 - 4 - 5 443 63
U22262.1 - 4 - 5 571 63
U22263.1 - 4 - 5 515 63
U22264.1 - 1 - 2 271 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003071.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK146427.1,AK151799.1,NM_001134391.1,NM_031159.3,U22262.1,U22263.1,U22264.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apobec1

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
response to osmotic stress
cytidine deamination
response to gamma radiation
cytidine to uridine editing
mRNA modification
regulation of cell proliferation
lipoprotein metabolic process
lipoprotein biosynthetic process
response to drug
lipoprotein transport
response to ethanol
mRNA stabilization
response to calcium ion
defense response to virus
DNA cytosine deamination
DNA demethylation
negative regulation of methylation-dependent chromatin silencing
catalytic activity
GTP cyclohydrolase activity
cytidine deaminase activity
cytosine deaminase activity
protein binding
tRNA-specific adenosine deaminase activity
zinc ion binding
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amidines
AU-rich element binding
N-cyclopropylmelamine deaminase activity
N-cyclopropylammeline deaminase activity
N-cyclopropylammelide alkylamino hydrolase activity
2,5-diamino-6-ribitylamino-4(3H)-pyrimidinone 5'-phosphate deaminase activity
tRNA-specific adenosine-37 deaminase activity
archaeal-specific GTP cyclohydrolase activity
metal ion binding
tRNA-specific adenosine-34 deaminase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099560 U22262.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099560 U22263.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099560 U22264.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099560 BC003792.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099560 NM_031159.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500430 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715074 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651105 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815729 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213508 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270178 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567979 AK048452.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335310 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96725 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887904 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558187 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538543 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406278 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517537 AF503920.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517537 BC064744.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517537 NM_144934.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546382 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685002 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46470 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 70008 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162318 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143836 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099560 U22262.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099560 U22263.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099560 U22264.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099560 BC003792.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099560 NM_031159.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160754 U22262.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160754 U22263.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160754 U22264.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160754 AK165209.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P160754 BC003792.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P55029 U22262.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P55029 U22263.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P55029 U22264.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P55029 NM_031159.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P55029 AK146427.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809551 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809558 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809552 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809550 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809560 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809555 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24809554 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162318 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01435860 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143836 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06052620 - -
Whole Mouse Genome G4122A A_51_P160754 U22262.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P160754 U22263.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P160754 U22264.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P160754 AK165209.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P160754 BC003792.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P55029 U22262.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P55029 U22263.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P55029 U22264.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P55029 NM_031159.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P55029 AK146427.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109522 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162318 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143836 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05021273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809497 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809395 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809446 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809546 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809555 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809468 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809543 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809592 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809454 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809544 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809509 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809474 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809473 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809465 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809550 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809576 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809415 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809466 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809483 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24809501 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809558 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05780015 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809410 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05481516 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809544 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809412 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809486 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809550 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809530 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809547 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809491 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809492 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809500 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809548 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809543 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809480 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05635137 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05569384 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05621879 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809537 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809487 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809494 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809552 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05127636 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05202473 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809499 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809560 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05202468 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809478 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809411 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05833561 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809539 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05222672 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809538 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809554 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809484 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809490 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05780018 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809408 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809531 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809551 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809415 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809498 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24809477 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10547627 - -
Affymetrix Mouse Gene 1.0 ST Array 10547629 - -
Affymetrix Mouse Gene 1.0 ST Array 10547623 - -
Affymetrix Mouse Gene 1.0 ST Array 10547630 - -
Affymetrix Mouse Gene 1.0 ST Array 10547626 - -
Affymetrix Mouse Gene 1.0 ST Array 10547625 - -
Affymetrix Mouse Gene 1.0 ST Array 10547631 - -
Affymetrix Mouse Gene 1.0 ST Array 10547622 - -
Affymetrix Mouse Gene 1.0 ST Array 10547632 - -
Affymetrix Mouse Gene 1.0 ST Array 10547624 - -
Affymetrix HT MG-430 PM Array Plate 1451755_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10547621 - -
Mouse Genome 430 2.0 Array 1451755_PM_a_at - -
Mouse Genome 430 2.0 Array 1451755_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11810_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11810_at - -
Affymetrix Mouse Clariom S Array PSR0600023787.mm.1 TC0600003071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600023787.mm.1 NM_001134391.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600023787.mm.1 NM_001134391.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600023787.mm.1 NM_031159.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600023787.mm.1 NM_031159.3 Refseq
Mouse Expression Set 430 1451755_a_at - -
Mouse Expression Set 430 1451755_PM_a_at - -
Murine Genome U74v2 Set 98398_s_at - -
Murine Genome U74v2 Set 98398_PM_s_at - -
Mouse Genome 430A 2.0 Array 1451755_a_at - -
Mouse Genome 430A 2.0 Array 1451755_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1451755_a_at - -
Affymetrix HT Mouse Genome 430A Array 1451755_PM_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 911726 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 4610022 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004610022 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257631 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1257631 - -
mouse-6 v1.1 (Illumina) scl28441.10_311-S - -
Illumina Sentrix mouse-6.v1 bead array 4610022 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl28441.10_311-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4610022 - -
MouseRef-8 v2 Expression BeadChip ILMN_1257631 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257631 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0