This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00483025_g1
See other CAR4 GE Assays ›
Gene Symbol:
Car4
Gene Name:
carbonic anhydrase 4
Gene Aliases:
AW456718, Ca4
Chromosome Location:
Chr.11: 84957754 - 84966054 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00483025_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Car4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007607.2 NP_031633.1 5 - 6 626 61
GenBank mRNA AK154754.1 - 5 - 6 592 61
AK165399.1 - 5 - 6 554 61
BC012704.1 - 5 - 6 551 4217743 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001285.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Car4

Gene Ontology Categories:

Function(s) Process(es)

one-carbon metabolic process
regulation of pH
carbon dioxide transport
bicarbonate transport
carbonate dehydratase activity
protein binding
zinc ion binding
lyase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100012161.mm.1 NM_007607.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100012161.mm.1 NM_007607.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100012161.mm.1 TC1100001285.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100012162.mm.1 NM_007607.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100012162.mm.1 NM_007607.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100012162.mm.1 TC1100001285.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline