This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00483112_m1
See other CCT5 GE Assays ›
Gene Symbol:
Cct5
Gene Name:
chaperonin containing Tcp1, subunit 5 (epsilon)
Gene Aliases:
Ccte, TCPE, mKIAA0098
Chromosome Location:
Chr.15: 31590813 - 31601804 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00483112_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007637.2 NP_031663.1 4 - 5 623 96
GenBank mRNA AK077753.1 - 4 - 5 581 96
AK088184.1 - 4 - 5 557 96
AK129056.1 - 4 - 5 529 96
AK133857.1 - 4 - 5 581 96
AK136493.1 - 4 - 5 575 96
AK145445.1 - 4 - 5 592 96
AK146405.1 - 4 - 5 595 96
AK146425.1 - 4 - 5 575 96
AK150390.1 - 4 - 5 542 96
AK153911.1 - 4 - 5 599 96
AK167895.1 - 4 - 5 564 96
BC092233.1 - 4 - 5 593 96
BC094427.1 - 4 - 5 570 30537966 96
Z31555.1 - 4 - 5 623 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001326.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct5

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
toxin transport
nucleotide binding
protein binding
ATP binding
G-protein beta-subunit binding
beta-tubulin binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500010298.mm.1 NM_007637.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500010298.mm.1 TC1500001326.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0