This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00483124_mH
See other CCT7 GE Assays ›
Gene Symbol:
Cct7
Gene Name:
chaperonin containing Tcp1, subunit 7 (eta)
Gene Aliases:
AA408524, AL022769, Ccth, Cctz
Chromosome Location:
Chr.6: 85451505 - 85468477 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00483124_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007638.4 NP_031664.3 7 - 8 1397 102
GenBank mRNA AK012201.1 - 7 - 8 853 102
AK146902.1 - 7 - 8 837 102
AK150160.1 - 7 - 8 884 102
AK166888.1 - 7 - 8 840 102
AK167367.1 - 7 - 8 854 102
AK167827.1 - 7 - 8 833 102
AK167938.1 - 7 - 8 837 102
AK168087.1 - 7 - 8 912 102
AK168272.1 - 7 - 8 833 102
AK169426.1 - 7 - 8 884 102
BC008255.1 - 7 - 8 826 2648519 102
Z31399.1 - 6 - 7 782 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000963.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct7

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
toxin transport
nucleotide binding
protein binding
ATP binding
identical protein binding
unfolded protein binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605103 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605102 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 Z31399.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 AK169426.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 AK198159.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 AK202338.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 BC008255.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 Z31399.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 AK169426.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 AK198159.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 AK202338.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P197259 BC008255.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24342 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377336 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861071 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601401 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 429794 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355477 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585921 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491039 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796201 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150375 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106438 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104399 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 Z31399.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 AK169426.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 AK198159.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 AK202338.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 BC008255.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 Z31399.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 AK169426.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 AK198159.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 AK202338.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P197259 BC008255.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 Z31399.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 AK169426.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 AK198159.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 AK202338.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 BC008255.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 Z31399.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 AK169426.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 AK198159.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 AK202338.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197259 BC008255.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605091 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605087 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605107 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605109 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605063 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605090 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605082 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605122 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605111 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605095 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605088 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605106 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605068 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605120 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605066 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605098 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605094 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605073 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605092 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605079 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605070 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605085 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605093 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605102 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605060 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605078 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605103 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605116 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605065 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605117 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605100 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605064 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605075 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605076 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605080 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605104 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24605114 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106438 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104399 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05966693 - -
Whole Mouse Genome G4122A A_51_P197259 Z31399.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 AK169426.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 AK198159.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 AK202338.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 BC008255.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 Z31399.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 AK169426.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 AK198159.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 AK202338.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P197259 BC008255.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105536 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102036 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104399 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106438 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05419033 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05419024 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605102 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605103 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605116 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605192 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605165 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605126 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605111 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605168 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605104 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605207 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605127 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605095 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605177 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605205 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605198 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605196 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605189 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605174 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605103 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605183 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605147 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605098 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605199 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605144 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605154 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605172 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605124 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605188 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605109 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605203 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605194 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605102 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605149 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605212 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605135 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605144 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605137 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605143 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605151 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605100 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605114 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605109 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05419030 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05545449 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605116 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05419018 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05784617 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605131 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605120 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605129 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605146 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05413344 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605149 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605102 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05701745 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605111 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605147 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605124 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605139 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605127 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24605098 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05701744 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10539614 - -
Affymetrix Mouse Gene 1.0 ST Array 10539612 - -
Affymetrix Mouse Gene 1.0 ST Array 10539608 - -
Affymetrix Mouse Gene 1.0 ST Array 10539607 - -
Affymetrix Mouse Gene 1.0 ST Array 10539616 - -
Affymetrix Mouse Gene 1.0 ST Array 10539615 - -
Affymetrix Mouse Gene 1.0 ST Array 10539613 - -
Affymetrix Mouse Gene 1.0 ST Array 10539611 - -
Affymetrix Mouse Gene 1.0 ST Array 10539610 - -
Affymetrix Mouse Gene 1.0 ST Array 10539609 - -
Affymetrix HT MG-430 PM Array Plate 1415817_PM_s_at - -
Affymetrix HT MG-430 PM Array Plate 1415816_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10539606 - -
Mouse Genome 430 2.0 Array 1415817_s_at - -
Mouse Genome 430 2.0 Array 1415817_PM_s_at - -
Mouse Genome 430 2.0 Array 1415816_PM_at - -
Mouse Genome 430 2.0 Array 1415816_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12468_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12468_PM_at - -
Affymetrix Mouse Clariom S Array PSR0600007449.mm.1 NM_007638.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007449.mm.1 NM_007638.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600007449.mm.1 TC0600000963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600007448.mm.1 NM_007638.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007448.mm.1 NM_007638.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600007448.mm.1 TC0600000963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600007446.mm.1 NM_007638.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007446.mm.1 NM_007638.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600007446.mm.1 TC0600000963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600007445.mm.1 NM_007638.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007445.mm.1 NM_007638.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600007445.mm.1 TC0600000963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600007447.mm.1 NM_007638.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007447.mm.1 NM_007638.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600007447.mm.1 TC0600000963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600007450.mm.1 NM_007638.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007450.mm.1 NM_007638.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600007450.mm.1 TC0600000963.mm.1 MTA TC
Mouse Expression Set 430 1415816_at - -
Mouse Expression Set 430 1415816_PM_at - -
Mouse Expression Set 430 1415817_s_at - -
Mouse Expression Set 430 1415817_PM_s_at - -
Murine Genome U74v2 Set 160562_PM_at - -
Murine Genome U74v2 Set 160562_at - -
Mouse Genome 430A 2.0 Array 1415816_at - -
Mouse Genome 430A 2.0 Array 1415816_PM_at - -
Mouse Genome 430A 2.0 Array 1415817_s_at - -
Mouse Genome 430A 2.0 Array 1415817_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1415817_s_at - -
Affymetrix HT Mouse Genome 430A Array 1415817_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1415816_at - -
Affymetrix HT Mouse Genome 430A Array 1415816_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 6660091 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006660091 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233793 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1233793 - -
mouse-6 v1.1 (Illumina) scl29820.13_336-S - -
Illumina Sentrix mouse-6.v1 bead array 6660091 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl29820.13_336-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660091 - -
MouseRef-8 v2 Expression BeadChip ILMN_1233793 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233793 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline