This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00483201_g1
See other CDH16 GE Assays ›
Gene Symbol:
Cdh16
Gene Name:
cadherin 16
Gene Aliases:
-
Chromosome Location:
Chr.8: 104601915 - 104624396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00483201_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252627.1 NP_001239556.1 10 - 11 1532 68
NM_001252628.1 NP_001239557.1 11 - 12 1622 68
NM_007663.3 NP_031689.1 11 - 12 1622 68
GenBank mRNA AF016271.1 - 11 - 12 1490 68
AJ609635.1 - 11 - 12 1489 68
AK052636.1 - 11 - 12 1425 68
AK093905.1 - 11 - 12 1420 68
AK164546.1 - 10 - 11 1533 68
BC015251.1 - 11 - 12 1470 4235757 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002806.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh16

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800021532.mm.1 NM_001252627.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021532.mm.1 NM_001252627.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021532.mm.1 NM_001252628.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021532.mm.1 NM_001252628.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021532.mm.1 NM_007663.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800021532.mm.1 NM_007663.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800021532.mm.1 TC0800002806.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0