This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00483205_g1
See other CDH16 GE Assays ›
Gene Symbol:
Cdh16
Gene Name:
cadherin 16
Gene Aliases:
-
Chromosome Location:
Chr.8: 104601915 - 104624396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00483205_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252627.1 NP_001239556.1 14 - 15 2343 68
NM_001252628.1 NP_001239557.1 15 - 16 2433 68
NM_007663.3 NP_031689.1 15 - 16 2433 68
GenBank mRNA AB009292.1 - 1 - 2 68
AF016271.1 - 15 - 16 2301 68
AJ609635.1 - 15 - 16 2300 68
AK052636.1 - 15 - 16 2236 68
AK093905.1 - 15 - 16 2231 68
AK164546.1 - 14 - 15 2344 68
BC015251.1 - 15 - 16 2281 4235757 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh16

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
calcium ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0