This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00483208_m1
See other CDH2 GE Assays ›
Gene Symbol:
Cdh2
Gene Name:
cadherin 2
Gene Aliases:
CDHN, N-cadherin, Ncad
Chromosome Location:
Chr.18: 16588877 - 16809049 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00483208_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007664.4 NP_031690.3 1 - 2 713 84
GenBank mRNA AB008811.1 - 1 - 2 390 84
AK032826.1 - 1 - 2 153 84
AK087418.1 - 1 - 2 127 84
AK146981.1 - 1 - 2 910 84
BC022107.1 - 1 - 2 144 5100313 84
M31131.1 - 1 - 2 390 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001026.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh2

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
synapse assembly
regulation of signal transduction
glial cell differentiation
single organismal cell-cell adhesion
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
cell migration
regulation of myelination
regulation of protein localization
regulation of Rho protein signal transduction
positive regulation of MAPK cascade
cell-cell adhesion mediated by cadherin
blood vessel morphogenesis
regulation of axonogenesis
striated muscle cell differentiation
protein heterooligomerization
negative regulation of canonical Wnt signaling pathway
neuroligin clustering involved in postsynaptic membrane assembly
neuronal stem cell population maintenance
regulation of postsynaptic density protein 95 clustering
positive regulation of synaptic vesicle clustering
calcium ion binding
protein binding
beta-catenin binding
enzyme binding
protein kinase binding
protein phosphatase binding
protein complex binding
alpha-catenin binding
gamma-catenin binding
metal ion binding
nitric-oxide synthase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline