This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00483984_m1
See other CRAT GE Assays ›
Gene Symbol:
Crat
Gene Name:
carnitine acetyltransferase
Gene Aliases:
AW107812, CARAT
Chromosome Location:
Chr.2: 30400476 - 30415748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00483984_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007760.3 NP_031786.2 1 - 2 435 88
GenBank mRNA AK041431.1 - 1 - 2 209 89
AK132179.1 - 2 - 3 262 88
AK170740.1 - 1 - 2 221 88
AK218779.1 - 1 - 2 43 88
BC006668.1 - 1 - 2 182 3499169 88
X85983.1 - 1 - 2 150 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003271.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK132179.1,AK170740.1,BC006668.1,NM_007760.3,X85983.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crat

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
transport
carnitine metabolic process, CoA-linked
carnitine O-acetyltransferase activity
receptor binding
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200027698.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027698.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027698.mm.1 TC0200003271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027687.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027687.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027687.mm.1 TC0200003271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027699.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027699.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027699.mm.1 TC0200003271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027703.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027703.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027703.mm.1 TC0200003271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027704.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027704.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027704.mm.1 TC0200003271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027685.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027685.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027685.mm.1 TC0200003271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027705.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027705.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027705.mm.1 TC0200003271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200027697.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027697.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027697.mm.1 TC0200003271.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 803369 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0