This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00483986_m1
See other CRAT GE Assays ›
Gene Symbol:
Crat
Gene Name:
carnitine acetyltransferase
Gene Aliases:
AW107812, CARAT
Chromosome Location:
Chr.2: 30400476 - 30415748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00483986_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007760.3 NP_031786.2 3 - 4 819 72
GenBank mRNA AK041431.1 - 3 - 4 594 72
AK132179.1 - 4 - 5 646 72
AK170740.1 - 3 - 4 605 72
BC006668.1 - 3 - 4 566 3499169 72
X85983.1 - 3 - 4 537 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003271.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crat

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
transport
carnitine metabolic process, CoA-linked
carnitine O-acetyltransferase activity
receptor binding
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200027682.mm.1 NM_007760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200027682.mm.1 NM_007760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200027682.mm.1 TC0200003271.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bs78j:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0