This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00484584_g1
See other GAA GE Assays ›
Gene Symbol:
Gaa
Gene Name:
glucosidase, alpha, acid
Gene Aliases:
E430018M07Rik
Chromosome Location:
Chr.11: 119267967 - 119285698 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00484584_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gaa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159324.1 NP_001152796.1 5 - 6 1121 80
NM_008064.3 NP_032090.3 5 - 6 1211 80
GenBank mRNA AK052211.1 - 5 - 6 1192 80
AK088481.1 - 5 - 6 1187 80
AK139333.1 - 5 - 6 1159 80
AK140042.1 - 5 - 6 1178 80
AK141546.1 - 5 - 6 1194 80
AK145631.1 - 4 - 5 626 80
AK146538.1 - 5 - 6 1115 80
AK150970.1 - 5 - 6 1169 80
AK210560.1 - 3 - 4 355 80
BC010210.1 - 5 - 6 1175 3969127 80
U49351.1 - 5 - 6 1063 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001999.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gaa

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
diaphragm contraction
heart morphogenesis
carbohydrate metabolic process
glycogen metabolic process
glycogen catabolic process
striated muscle contraction
lysosome organization
locomotory behavior
metabolic process
tissue development
vacuolar sequestering
muscle cell cellular homeostasis
neuromuscular process controlling posture
neuromuscular process controlling balance
cardiac muscle contraction
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
alpha-1,4-glucosidase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
carbohydrate binding
maltose alpha-glucosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100019028.mm.1 NM_001159324.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100019028.mm.1 NM_008064.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100019028.mm.1 TC1100001999.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0