This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00484590_g1
See other GAA GE Assays ›
Gene Symbol:
Gaa
Gene Name:
glucosidase, alpha, acid
Gene Aliases:
E430018M07Rik
Chromosome Location:
Chr.11: 119267967 - 119285698 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00484590_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gaa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159324.1 NP_001152796.1 12 - 13 1927 57
NM_008064.3 NP_032090.3 12 - 13 2017 57
GenBank mRNA AK052211.1 - 12 - 13 1998 57
AK088481.1 - 12 - 13 1993 57
AK139333.1 - 12 - 13 1965 57
AK140042.1 - 12 - 13 1985 57
AK145631.1 - 11 - 12 1433 57
AK146538.1 - 12 - 13 1921 57
AK150970.1 - 12 - 13 1975 57
BC010210.1 - 12 - 13 1981 3969127 57
U49351.1 - 12 - 13 1869 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001999.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gaa

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
diaphragm contraction
heart morphogenesis
carbohydrate metabolic process
glycogen metabolic process
glycogen catabolic process
striated muscle contraction
lysosome organization
locomotory behavior
metabolic process
tissue development
vacuolar sequestering
muscle cell cellular homeostasis
neuromuscular process controlling posture
neuromuscular process controlling balance
cardiac muscle contraction
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
alpha-1,4-glucosidase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
carbohydrate binding
maltose alpha-glucosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100019039.mm.1 NM_001159324.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100019039.mm.1 NM_008064.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100019039.mm.1 TC1100001999.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100019038.mm.1 NM_001159324.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100019038.mm.1 NM_008064.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100019038.mm.1 TC1100001999.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0