This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00485041_m1
See other MME GE Assays ›
Gene Symbol:
Mme
Gene Name:
membrane metallo endopeptidase
Gene Aliases:
6030454K05Rik, C85356, CALLA, CD10, NEP, SFE
Chromosome Location:
Chr.3: 63295226 - 63383713 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00485041_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mme
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289462.1 NP_001276391.1 16 - 17 2056 95
NM_001289463.1 NP_001276392.1 16 - 17 1840 95
NM_008604.4 NP_032630.2 16 - 17 1690 95
GenBank mRNA AK031446.1 - 16 - 17 1691 95
AK033824.1 - 16 - 17 1751 95
BC034092.1 - 16 - 17 1712 4483558 95
BC066840.1 - 15 - 16 1599 95
M81591.1 - 15 - 16 1602 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000500.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mme

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
peptide metabolic process
sensory perception of pain
creatinine metabolic process
beta-amyloid metabolic process
cellular response to cytokine stimulus
cellular response to UV-A
cellular response to UV-B
endopeptidase activity
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
exopeptidase activity
zinc ion binding
hydrolase activity
peptide binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0