This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00485110_g1
See other MDH1 GE Assays ›
Gene Symbol:
Mdh1
Gene Name:
malate dehydrogenase 1, NAD (soluble)
Gene Aliases:
B230377B03Rik, D17921, MDH-s, MDHA, Mor-2, Mor2
Chromosome Location:
Chr.11: 21556692 - 21571934 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00485110_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mdh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008618.3 NP_032644.3 6 - 7 874 77
GenBank mRNA AK002404.1 - 6 - 7 732 77
AK005237.1 - 6 - 7 769 77
AK164785.1 - 6 - 7 755 77
AK168545.1 - 6 - 7 712 77
BC050940.1 - 6 - 7 740 6820601 77
M29462.1 - 6 - 7 753 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002329.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mdh1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
carbohydrate metabolic process
tricarboxylic acid cycle
oxaloacetate metabolic process
malate metabolic process
NADH metabolic process
NAD metabolic process
carboxylic acid metabolic process
oxidation-reduction process
mitophagy in response to mitochondrial depolarization
catalytic activity
oxidoreductase activity
malate dehydrogenase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
L-malate dehydrogenase activity
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100022099.mm.1 NM_008618.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100022099.mm.1 NM_008618.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100022099.mm.1 TC1100002329.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100022102.mm.1 NM_008618.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100022102.mm.1 NM_008618.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100022102.mm.1 TC1100002329.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline