Assay Details
| Gene Symbol: Mpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089180.1 | - | 4 - 5 | 399 | 65 | |
| AK157960.1 | - | 2 - 3 | 292 | 65 | ||
| BC139139.1 | - | 2 - 3 | 260 | 8862161 | 65 | |
| BC141226.1 | - | 2 - 3 | 260 | 9055838 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpz
Genomic Map