This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00485277_m1
See other MTHFD2 GE Assays ›
Gene Symbol:
Mthfd2
Gene Name:
methylenetetrahydrofolate dehydrogenase (NAD+ dependent), methenyltetrahydrofolate cyclohydrolase
Gene Aliases:
AW558851, NMDMC
Chromosome Location:
Chr.6: 83305704 - 83325947 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00485277_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mthfd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008638.2 NP_032664.1 2 - 3 359 82
GenBank mRNA AK076019.1 - 2 - 3 312 82
AK076801.1 - 2 - 3 309 82
AK152249.1 - 2 - 3 351 82
AK159680.1 - 2 - 3 332 82
AK165840.1 - 2 - 3 351 82
AK166262.1 - 2 - 3 342 82
BC019511.1 - 2 - 3 346 4164253 82
J04627.1 - 2 - 3 302 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002565.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mthfd2

Gene Ontology Categories:

Function(s) Process(es)

one-carbon metabolic process
metabolic process
folic acid-containing compound biosynthetic process
tetrahydrofolate metabolic process
oxidation-reduction process
magnesium ion binding
catalytic activity
methenyltetrahydrofolate cyclohydrolase activity
methylenetetrahydrofolate dehydrogenase (NAD+) activity
methylenetetrahydrofolate dehydrogenase (NADP+) activity
oxidoreductase activity
hydrolase activity
phosphate ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600019915.mm.1 NM_008638.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600019915.mm.1 NM_008638.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600019915.mm.1 TC0600002565.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline