This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00485637_g1
See other RBPJL GE Assays ›
Gene Symbol:
Rbpjl
Gene Name:
recombination signal binding protein for immunoglobulin kappa J region-like
Gene Aliases:
RBP-L, Rbpsuhl
Chromosome Location:
Chr.2: 164403056 - 164415448 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00485637_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbpjl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009036.1 NP_033062.1 8 - 9 964 60
GenBank mRNA AK052449.1 - 6 - 7 4075 60
AK131814.1 - 8 - 9 945 60
BC152311.1 - 8 - 9 958 40090714 60
Y10926.1 - 8 - 9 964 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002515.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbpjl

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200020508.mm.1 NM_009036.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020508.mm.1 NM_009036.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020508.mm.1 TC0200002515.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0