This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00485794_m1
See other RPIA GE Assays ›
Gene Symbol:
Rpia
Gene Name:
ribose 5-phosphate isomerase A
Gene Aliases:
RPI
Chromosome Location:
Chr.6: 70765720 - 70792175 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00485794_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpia
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009075.2 NP_033101.2 5 - 6 534 76
GenBank mRNA AK137235.1 - 5 - 6 369 76
BC053526.1 - 5 - 6 357 76
L35034.1 - 5 - 6 514 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002452.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rpia

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

pentose-phosphate shunt
pentose-phosphate shunt, non-oxidative branch
ribose phosphate metabolic process
ribose-5-phosphate isomerase activity
isomerase activity
carbohydrate binding
monosaccharide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600018975.mm.1 NM_009075.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018975.mm.1 NM_009075.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600018975.mm.1 TC0600002452.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600018973.mm.1 NM_009075.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600018973.mm.1 NM_009075.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600018973.mm.1 TC0600002452.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline