This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486082_m1
See other SGPL1 GE Assays ›
Gene Symbol:
Sgpl1
Gene Name:
sphingosine phosphate lyase 1
Gene Aliases:
AI428538, D10Xrf456, S1PL, Spl
Chromosome Location:
Chr.10: 61098642 - 61147686 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486082_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sgpl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009163.3 NP_033189.2 6 - 7 679 98
GenBank mRNA AF036894.1 - 5 - 6 549 98
AK036747.1 - 6 - 7 567 98
AK037789.1 - 6 - 7 564 98
AK043024.1 - 6 - 7 566 98
AK049342.1 - 6 - 7 587 98
AK150053.1 - 6 - 7 574 98
AK173137.1 - 6 - 7 608 98
BC026135.1 - 6 - 7 625 4212293 98
CT010214.1 - 5 - 6 489 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sgpl1

Gene Ontology Categories:

Function(s) Process(es)

luteinization
vasculogenesis
kidney development
lipid metabolic process
fatty acid metabolic process
sphingolipid metabolic process
ceramide metabolic process
nitrogen compound metabolic process
apoptotic process
spermatogenesis
androgen metabolic process
estrogen metabolic process
female gonad development
post-embryonic development
fibroblast migration
carboxylic acid metabolic process
hemopoiesis
sphingolipid catabolic process
Leydig cell differentiation
regulation of multicellular organism growth
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
palate development
face morphogenesis
apoptotic signaling pathway
catalytic activity
sphinganine-1-phosphate aldolase activity
lyase activity
carboxy-lyase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0