This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486113_m1
See other SIAH1B GE Assays ›
Gene Symbol:
Siah1b
Gene Name:
seven in absentia 1B
Gene Aliases:
AA960570, Sinh1b, siah-1b
Chromosome Location:
Chr.X: 164070703 - 164076664 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486113_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Siah1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC052887.1 - 3 - 4 189 30053109 97
CF723667.1 - 3 - 4 445 97
CJ063158.1 - 3 - 4 266 97
Z19580.1 - 3 - 4 194 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00003329.mm.1

Additional Information:

This assay is in the 5'UTR region of the BC052887.1,Z19580.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796354 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796349 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796353 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796348 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796351 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796350 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796347 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796352 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972182 Z19580.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972182 NM_009173.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972182 BC052887.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400426 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530406 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657973 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833960 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138399 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152668 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972182 Z19580.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972182 NM_009173.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972182 BC052887.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P378602 Z19580.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P378602 Z19579.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P378602 NM_009173.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P378602 BC052887.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P378602 NM_009172.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P44994 Z19580.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P44994 NM_009173.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P44994 BC052887.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796341 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796338 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796356 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796360 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796378 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796368 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796339 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796358 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796337 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796344 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796371 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796359 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796395 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796349 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796336 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796362 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796350 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796382 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796380 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796345 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796370 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796365 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796367 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796342 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796373 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796396 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796355 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796376 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796334 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796335 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796398 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796363 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796374 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796352 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796348 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32796347 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152668 - -
Whole Mouse Genome G4122A A_51_P378602 Z19580.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P378602 Z19579.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P378602 NM_009173.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P378602 BC052887.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P378602 NM_009172.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P44994 Z19580.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P44994 NM_009173.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P44994 BC052887.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113377 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152668 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14918305 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14918316 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14918312 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14918310 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796353 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796349 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796350 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796347 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796348 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796354 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796351 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32796352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796258 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796340 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796306 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796323 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796292 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796270 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796332 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796287 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32796371 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796344 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796323 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796371 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14680828 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796370 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796380 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796301 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796306 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796360 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796367 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14918318 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14918302 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796322 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796382 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796352 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796327 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796383 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14625182 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796309 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14617650 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796373 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796326 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796339 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796386 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796335 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14918321 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796365 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796333 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796385 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796374 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796337 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796362 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14918316 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796300 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796348 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796329 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796341 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796312 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796330 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796338 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796368 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796336 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32796378 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14518297 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10607749 - -
Affymetrix Mouse Gene 1.0 ST Array 10607748 - -
Affymetrix Mouse Gene 1.0 ST Array 10607750 - -
Affymetrix Mouse Gene 1.0 ST Array 10607751 - -
Affymetrix Mouse Gene 1.1 ST Array 10607747 - -
Mouse Genome 430 2.0 Array 1419404_s_at - -
Mouse Genome 430 2.0 Array 1419404_PM_s_at - -
Affymetrix Mouse Clariom S Array PSR0X00020739.mm.1 NM_009173.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020739.mm.1 TC0X00003329.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00020738.mm.1 NM_009173.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020738.mm.1 TC0X00003329.mm.1 MTA TC
Mouse Expression Set 430 1419404_PM_s_at - -
Mouse Expression Set 430 1419404_s_at - -
Murine Genome U74v2 Set 92290_at - -
Murine Genome U74v2 Set 92290_PM_at - -
Mouse Genome 430A 2.0 Array 1419404_s_at - -
Mouse Genome 430A 2.0 Array 1419404_PM_s_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6290519 - -
MouseWG-6 v2 Expression BeadChip 1170309 - -
MouseWG-6 v2 Expression BeadChip 6200520 - -
Illumina mouse-6 v1.0 expression beadchip 6290519 - -
Illumina mouse-6 v1.0 expression beadchip 6200520 - -
Illumina mouse-6 v1.0 expression beadchip 4050279 - -
Illumina mouse-6 v1.0 expression beadchip 1170309 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006290519 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001170309 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004050279 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006200520 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618014 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217391 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646574 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2706393 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1217391 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2618014 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2646574 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2706393 - -
mouse-6 v1.1 (Illumina) scl53613.6.1_106-S - -
mouse-6 v1.1 (Illumina) scl020438.3_0-S - -
mouse-6 v1.1 (Illumina) scl0020438.2_182-S - -
mouse-6 v1.1 (Illumina) scl0020438.2_18-S - -
Illumina Sentrix mouse-6.v1 bead array 4050279 - -
Illumina Sentrix mouse-6.v1 bead array 6290519 - -
Illumina Sentrix mouse-6.v1 bead array 1170309 - -
Illumina Sentrix mouse-6.v1 bead array 6200520 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl020438.3_0-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0020438.2_182-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl53613.6.1_106-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0020438.2_18-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050279 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6200520 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290519 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170309 - -
MouseRef-8 v2 Expression BeadChip ILMN_2706393 - -
MouseRef-8 v2 Expression BeadChip ILMN_2646574 - -
MouseRef-8 v2 Expression BeadChip ILMN_2903734 - -
MouseRef-8 v2 Expression BeadChip ILMN_2618014 - -
MouseRef-8 v2 Expression BeadChip ILMN_1217391 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217391 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2706393 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2618014 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2903734 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646574 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0