This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486288_m1
See other SOAT1 GE Assays ›
Gene Symbol:
Soat1
Gene Name:
sterol O-acyltransferase 1
Gene Aliases:
8430426K15Rik, ACAT-1, AW550831, Acact, ald, hid
Chromosome Location:
Chr.1: 156428108 - 156474328 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486288_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Soat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009230.3 NP_033256.2 13 - 14 1494 70
GenBank mRNA AK052299.1 - 10 - 11 1051 70
AK142486.1 - 13 - 14 1411 70
AK159393.1 - 13 - 14 1424 70
AK159529.1 - 13 - 14 1424 70
AK160017.1 - 13 - 14 1424 70
BC083092.1 - 12 - 13 1308 30446741 70
DQ903181.1 - 13 - 14 1411 70
S81092.1 - 13 - 14 1428 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Soat1

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
steroid metabolic process
cholesterol metabolic process
macrophage derived foam cell differentiation
cholesterol storage
cholesterol efflux
very-low-density lipoprotein particle assembly
cholesterol esterification
cholesterol homeostasis
positive regulation of amyloid precursor protein biosynthetic process
fatty-acyl-CoA binding
sterol O-acyltransferase activity
O-acyltransferase activity
cholesterol binding
transferase activity
transferase activity, transferring acyl groups
cholesterol O-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0